Order Entry
United Kingdom
ContactUsLinkComponent
 
Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®
 :  Merck
 
Platinum 10% on activated carbon hydrogenation catalyst, for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Environmentally Preferable:

 

 

  • Solubility:
    (20 °C) insoluble
  • Bulk Density:
    250 kg/m3
  • pH:
    2 - 10 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    600 °C

 

 

Frequently Bought Together