Calcium chloride, ultra dry ≥99.9% (metals basis)
: Chloro calcium, Calcium dichloride, CaCl2
:10043-52-4
:
:CaCl₂
:MFCD00010903
:>1600 °C
:782 °C
:2.15
:110.98 g/mol
:14,1659
:Ampuled under argon
:Ambient
:233-140-8
:01-2119494219-28
:GHS07
- Environmentally Preferable:
Frequently Bought Together