Order Entry
United Kingdom
ContactUsLinkComponent
 
Water, Ultrapure for HPLC
    
 
Water, Ultrapure for HPLC
    
 

 

  • Environmentally Preferable:

 

 

  • Appearance (Color):
    Clear colorless
  • Form:
    Liquid
  • Assay (unspecified):
    Specific Conductance at 25°C and time packaging: 1.5 microS/cm, max
  • Optical Absorbance at:
    400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max
  • Residue on Evaporation:
    2 ppm, Max

 

Frequently Bought Together