Potassium bromide ≥99.5%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7758-02-3
:
Formula:KBr
MDL Number:MFCD00011358
Boiling Pt:1435
Melting Pt:730
Density:2.75 g/cm³ (20 °C)
Molecular Weight:119 g/mol
Merck Index:13,07701
EINECS:231-830-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 99.5 %
- Acidity or alkalinity:Passes test
- pH (20°C; 5 %):5 - 8
- Heavy metals (as Pb):Max. 5 ppm
- Insolubility in water:Max. 50 ppm
- Loss on drying (105°C):Max. 0.5 %
- BrO3 (Bromate):Max. 10 ppm
- Cl (Chloride):Max. 0.1 %
- I (Iodide):Max. 10 ppm
- IO3 (Iodate):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ba (Barium):Max. 20 ppm
- Ca (Calcium):Max. 10 ppm
- Fe (Iron):Max. 5 ppm
- Mg (Magnesium):Max. 10 ppm
- Na (Sodium):Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
26708_POTASSIUM BROMIDE_BSE_REV0
BSE/TSE Certificates
spec_VWRC26708.238 spec_VWRC26708.295 spec_VWRC26708.295
Product specification
Frequently Bought Together