cis-Dichlorobis(triphenylphosphite)platinum(II) (≥24,2% Pt)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: cis-Bis(triphenylphosphine)palladium (II) chloride, cis-Bis(triphenylphosphine)platinum(II) chloride, Pt(PPh₃)₂Cl₂
CAS Number:15604-36-1
:30053-58-8
Formula:[(C₆H₅)₃P]₂PtCl₂
MDL Number:MFCD00010825
Molecular Weight:790.56 g/mol
Melting Pt:310 °C (dec.)
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together