Disposal bin, SHARPSGUARD®, yellow
About this item
SHARPSGUARD® yellow is a similar range of containers to SHARPSGUARD® orange, with the added advantage that they can also be used to dispose of sharps contaminated with medicinal products and their residues other than cytotoxic and/or cytostatic medicines. To ensure the containers are safe and simple to use the opaque doors clearly show when the door is in the temporary or fully closed position. The SHARPSGUARD® yellow range is designed to be incinerated or disposed of by other authorised process.
- PP
The SHARPSGUARD® yellow theatre bin (129-1047) is designed to be used upright to ease the disposal of long instruments such as staple guns as well as chest drains. It is ideal for theatres, acute medical areas and X-ray departments but is not recommended for high public access areas.
For the SHARPSGUARD® yellow 22 XA bin (129-0722) the 'XA' denotes extra access. This container is designed for the disposal of larger instruments. It has a hands-free hinged door, which can stay open at 45 and 90 degrees, providing a large aperture for the disposal of large instruments with minimum handling.
Certifications: They are manufactured to the most stringent quality and safety standards: BS EN ISO 23907, UN3291, AFNOR NFX 30-500 and ISO 9002.
4 Options Available Below
Product Details & Documents
SHARPSGUARD® yellow is a similar range of containers to SHARPSGUARD® orange, with the added advantage that they can also be used to dispose of sharps contaminated with medicinal products and their residues other than cytotoxic and/or cytostatic medicines. To ensure the containers are safe and simple to use the opaque doors clearly show when the door is in the temporary or fully closed position. The SHARPSGUARD® yellow range is designed to be incinerated or disposed of by other authorised process.
- PP
The SHARPSGUARD® yellow theatre bin (129-1047) is designed to be used upright to ease the disposal of long instruments such as staple guns as well as chest drains. It is ideal for theatres, acute medical areas and X-ray departments but is not recommended for high public access areas.
For the SHARPSGUARD® yellow 22 XA bin (129-0722) the 'XA' denotes extra access. This container is designed for the disposal of larger instruments. It has a hands-free hinged door, which can stay open at 45 and 90 degrees, providing a large aperture for the disposal of large instruments with minimum handling.
Certifications: They are manufactured to the most stringent quality and safety standards: BS EN ISO 23907, UN3291, AFNOR NFX 30-500 and ISO 9002.
Specifications for Product Family
Specifications
- Catalog No:
- DANIDD442NRYL
- 129-1063
- 129-1041
- 129-1043
- Material:
- PP
- PP
- PP
- PP
Specifications
Product Family Options
Product Information
- CapacityColourW×D×HØ×HAvailabilityPrice
- DANIDD442NRYLSHARPSGUARD® yellow 0.5SHARPSGUARD® yellow 0.50, 5 LYellow/yellow43×80×200 mm- mmLog in for availability1 * 60 items£153.00Specifications:
More
Specifications
Cat. No.DANIDD442NRYLMaterialPP - 129-1063SHARPSGUARD® yellow 1Specifications:
More
Specifications
Cat. No.129-1063MaterialPP - 129-1041SHARPSGUARD® yellow 7Specifications:
More
Specifications
Cat. No.129-1041MaterialPP - 129-1043SHARPSGUARD® yellow 22Specifications:
More
Specifications
Cat. No.129-1043MaterialPP