2,4,6-Trimethylaniline 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:88-05-1
:
Formula:C₉H₁₃N
MDL Number:MFCD00007740
Boiling Pt:233 °C (760 mmHg)
Melting Pt:–5 °C
Density:0.96 g/cm³
Molecular Weight:135.21 g/mol
Flash Pt:96 °C
Storage Temperature:Ambient
EINECS:201-794-3
UN:2810
ADR:6.1 III
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5505 to 1.5525 (20°C, 589 nm)
- GC:≥96.0 %
- Appearance of solution:Clear colorless
- Residue after evaporation:≤0.05 %
- Related substances:≤0.15 % (Benzaldehyde)
- Acidity:Passes test
- Peroxide value:≤5
Specification Test Results
Frequently Bought Together