α-Chloro-o-xylene 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Methylbenzyl chloride
CAS Number:552-45-4
:
Formula:C₈H₉Cl
MDL Number:MFCD00000899
Boiling Pt:197…199 °C (760 mmHg)
Melting Pt:–2 °C
Density:1.08 g/cm³
Molecular Weight:140.61 g/mol
Flash Pt:73 °C
UN:3265
ADR:8 II
Merck Index:12,10224
EINECS:209-013-8
:GHS07
:GHS05
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5405 to 1.5425 (20°C, 589 nm)
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together