Tribenzylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:620-40-6
:
Formula:N(CH₂C₆H₅)₃
MDL Number:MFCD00004773
Boiling Pt:380
Melting Pt:92
Density:0.9912 g/cm³ (95 °C)
Molecular Weight:287.4 g/mol
Flash Pt:65 °C (closed cup)
Storage Temperature:Ambient
EINECS:210-638-3
UN:1325
ADR:4.1 III
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together