2,3-Difluoro-4-ethoxyphenylboronic acid
Supplier: COMBI-BLOCKS
Certificates
About this item
Synonyms: 2,3-Difluoro-4-ethoxybenzeneboronic acid, 4-Ethoxy-2,3-difluorobenzeneboronic acid, 4-Ethoxy-2,3-difluorophenylboronic acid, (4-Ethoxy-2,3-difluorophenyl)boronic acid
CAS Number:212386-71-5
:
Formula:C₈H₉BF₂O₃
MDL Number:MFCD09258743
Molecular Weight:201.96 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together