3-Phenylbenzoic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid
CAS Number:716-76-7
:
Formula:C₁₃H₁₀O₂
MDL Number:MFCD00045846
Molecular Weight:198.22 g/mol
Melting Pt:164…169 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Off-white to beige Crystalline powder
- Infrared spectrum:Conforms
- Melting point:164 to 169 °C
- Titration with NaOH:≥96.0 %
Specification Test Results
Frequently Bought Together