Order Entry
United Kingdom
ContactUsLinkComponent
Syringe filters, Minisart® HY
Syringe filters, Minisart® HY
Supplier:  Sartorius
Certificates
undefined
Syringe filters, Minisart® HY
Supplier:  Sartorius
Certificates
Specifications
  • Catalog No:
  • 514-0676
  • 512-3222
  • 611-0696
  • Airflow rate:
  • 2000 ml/min with air at 0,1 bar (1,5 psi)
  • 2000 ml/min with air at 0,1 bar (1,5 psi)
  • 2000 ml/min with air at 0,1 bar (1,5 psi)
  • Max. operating pressure:
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • 4,5 bar (65 psi)
  • Max. temperature:
  • 60 °C (140 °F)
  • 60 °C (140 °F)
  • 60 °C (140 °F)