About this item
The F-series chillers provide the final step to a fully integrated evaporation or distillation process, leading to several key benefits:
- Up to 75% less energy consumption
- Reduced heat emission
- Easy handling and mobility
- Optimised distillation capacity
- Increased process safety due to communication of all connected instruments
F-100: Cost effective, small recirculating chiller that fulfils the basic cooling requirements. The perfect replacement of tap water chilling with small distillation instruments. It cools to a fixed temperature of 10 °C and is ideal for use with one rotary evaporator.
F-108: Powerful alternative to the F-100. The chiller is capable of cooling to −10 °C. At 15 °C the F-108 provides 800 W cooling capacity. If connected to one rotary evaporator it is possible to keep the temperature at 0 °C when performing a normal distillation.
F-114: Ideal when very low temperatures are required for single or multiple units. At 15 °C the F-114 provides a powerful 1400 W cooling capacity and ultimately cools to a final temperature of −10 °C.
F-125: A powerful industrial chiller with a cooling capacity of 2500 W at 15 °C and an integrated trolley for use with the Rotavapor® R-220 SE industrial rotary evaporator.
F-105: is environmentally friendly and compact recirculation chiller designed for one laboratory evaporator. This features an adjustable temperature with a higher cooling capacity. In addition, the F-105 offers ON/OFF regulation when used in conjunction with the Interface. Cooling capacity of 530 W at 15 °C, 50/60 Hz frequency.
Specifications
- Catalog No:
- 531-0858
- Model:
- F-100
- Cooling power:
- 500 W