Salicylic acid 99+% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid
CAS Number:69-72-7
:
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439
Boiling Pt:211 °C (20 mmHg)
Melting Pt:158…160 °C
Density:1.443
Molecular Weight:138.12 g/mol
Flash Pt:157 °C (315 °F)
Merck Index:14,8332
Storage Temperature:Ambient
EINECS:200-712-3
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together