Bossheads, globular
Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
- 35 mm Ø
- For rods up to 13 mm Ø
- Nickel plated hinged screws
Weight: 100 g
Frequently Bought Together
Bossheads made of zinc die-cast, either natural, powder coated or nickel plated.
Weight: 100 g