3-Nitrobenzonitrile 98%
: m-Nitrobenzonitrile
:619-24-9
:
:C₇H₄N₂O₂
:MFCD00007194
:165 °C (21 mmHg)
:148.12 g/mol
:115…119 °C
:Ambient
:210-587-7
:3439
:6.1 II
:GHS06
- Environmentally Preferable:
- Appearance:Yellow Crystalline powder or needles
- Infrared spectrum:Conforms
- Melting point:115 to 119 °C
- GC:≥97.5 %
Frequently Bought Together