Order Entry
United Kingdom
ContactUsLinkComponent
Low TOC cleaning validation swabs, Alpha®
 :  Texwipe
undefined
Low TOC cleaning validation swabs, Alpha®
 :  Texwipe
  • Catalog No:
  • 115-1898
  • 115-1894
  • Model:
  • TX761K
  • TX714K
  • Head length:
  • 17 mm
  • 26 mm
  • Head width:
  • 7 mm
  • 13 mm
  • Handle length:
  • 145 mm
  • 102 mm
  • Handle width:
  • 3,2 mm
  • 5 mm
Frequently Bought Together