Vanadyl(IV) acetylacetonate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)
CAS Number:3153-26-2
:
Formula:C₁₀H₁₄O₅V
MDL Number:MFCD00000032
Molecular Weight:265.16 g/mol
ADR:6.1 III
:M-Clarity™ quality level = MQ100
EINECS:221-590-8
UN:3285
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together