Vanadyl(IV) acetylacetonate
: Vanadium(IV) oxide acetylacetonate, Oxobis(2,4-pentanedionato-O,O')vanadium (IV), Vanadium(IV) bis(acetylacetonato)oxide, Vanadyl acetylacetonate, Di(2,4-pentandionato-O,O')vanadium(IV)oxid, Vanadium (IV) oxide bis(2,4-pentanedionate)
:3153-26-2
:
:C₁₀H₁₄O₅V
:MFCD00000032
:140 °C (13 mmHg)
:256…259 °C
:1.4
:265.16 g/mol
:Ambient
:221-590-8
:3285
:6.1 III
:GHS06
:GHS07
- Environmentally Preferable:
Frequently Bought Together