Disodium hydrogen citrate sesquihydrate 99%, pure
: Citric acid disodium salt sesquihydrate, Sodium hydrogencitrate sesquihydrate, Disodium hydrogen 2-hydroxypropane-1,2,3-tricarboxylate sesquihydrate, Sodium citrate dibasic sesquihydrate
:6132-05-4
:
:C₆H₆Na₂O₇·1.5H₂O
:MFCD00150445
:263.11 g/mol
:149 °C (dec.)
:Ambient
:242-734-6
- Environmentally Preferable:
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- pH:4.9 to 5.2 (3 % solution)
- Titration with HClO4:98.5 to 104.0 %
- Insoluble matter:≤0.01 % (in water)
Frequently Bought Together