9-Methylanthracene for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:779-02-2
:
Formula:C₁₅H₁₂
MDL Number:MFCD00001261
Boiling Pt:347
Melting Pt:73…76 °C
Density:1.06 g/cm³ (20 °C)
Molecular Weight:192.26 g/mol
EINECS:212-299-7
ADR:9 III
:M-Clarity™ quality level = MQ200
UN:3077
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:0.0003 mg/l (25 °C)
- Melting Point:76 - 79 °C
- Molar Mass:192.26 g/mol
- Boiling Point:196 - 197 °C (16 hPa)
- Refractive Index:1.6959 (99 °C, 589 nm)
- Density:1.06 g/cm3
Specification Test Results
Frequently Bought Together