1-Pentene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: alpha-n-Amylene, Propylethylene
CAS Number:109-67-1
:
Formula:C₅H₁₀
MDL Number:MFCD00003567
Boiling Pt:30
Melting Pt:–165 °C
Density:0.641 g/cm³ (20 °C)
Molecular Weight:70.13 g/mol
Flash Pt:–20 °C
Storage Temperature:Refrigerator
UN:1108
ADR:3 I
Merck Index:12,07262
EINECS:203-694-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together