2,3-Dimethylbutane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:79-29-8
:
Formula:C₆H₁₄
MDL Number:MFCD00008925
Boiling Pt:58
Melting Pt:–128.5 °C
Density:0.6615 g/cm³ (20 °C)
Molecular Weight:86.18 g/mol
EINECS:201-193-6
UN:2457
ADR:3 II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together