About this item
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:151 °C (1013 hPa)
- Melting Pt:–53 °C
- Density:0.718 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- ADR:3,III
- :M-Clarity™ quality level = MQ200
- UN:1920
3 Options Available Below
Product Details & Documents
- CAS Number:111-84-2
- Formula:H₃C(CH₂)₇CH₃
- MDL Number:MFCD00009574
- Molecular Weight:128.26 g/mol
- Boiling Pt:151 °C (1013 hPa)
- Melting Pt:–53 °C
- Density:0.718 g/cm³ (20 °C)
- Flash Pt:31 °C
- Storage Temperature:Ambient
- EINECS:203-913-4
- ADR:3,III
- :M-Clarity™ quality level = MQ200
- UN:1920
Documents
msds_806838
SDS
ProductPage_806838
Product specification
Specifications for Product Family
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-53 °C
- Molar Mass:128.25 g/mol
- Boiling Point:151 °C
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:31 °C
- Refractive Index:- 7 mm2/s (40 °C)
- Explosion Limit:0.7 - 5.6 %(V)
- Density:0.72 g/cm3 (20 °C)
- Ignition Point:205 °C
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



