Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
Density:1.1 g/cm³ (20 °C)
Molecular Weight:288.38 g/mol
Melting Pt:204…207 °C
Flash Pt:>100 °C
ADR:4.1
Merck Index:14,08636
:M-Clarity™ quality level = MQ100
EINECS:205-788-1
UN:1325
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together