1,4-Butanediol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1,4-Dihydroxy butane, BDO, Tetramethylene glycol, 1,4-Butylene glycol
CAS Number:110-63-4
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002968
Boiling Pt:230
Melting Pt:19.5…20 °C
Density:1.017 g/cm³ (20 °C)
Molecular Weight:90.12 g/mol
:M-Clarity™ quality level = MQ200
EINECS:203-786-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:19.5 - 20.0 °C
- Molar Mass:90.12 g/mol
- Boiling Point:230 °C (1013 hPa)
- Vapor Pressure:- 1 hPa (20 °C)
- Flash Point:134 °C
- Refractive Index:1.4446 (25 °C, 589 nm)
- Explosion Limit:2.4 - 15.3 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:7 - 8 (500 g/l, H2O, 20 °C)
- Ignition Point:385 °C
Specification Test Results
Related Information
Frequently Bought Together