Ethanol absolute ≥99.7% for molecular biology DNase-, RNase-free (not denatured)
Catalog # SERA39556.02
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ethyl alcohol
:
:46.07 g/mol
Boiling Pt:78.3
Flash Pt:12 °C (closed cup)
EINECS:200-578-6
UN:1170
:GHS02
Storage Temperature:Ambient
CAS Number:64-17-5
Formula:C₂H₅OH
MDL Number:MFCD00003568
Melting Pt:–114.5 °C
Density:0.79 g/cm³ (20 °C)
ADR:3 II
Merck Index:12,03806