Order Entry
United Kingdom
ContactUsLinkComponent
 
Ethanol absolute ≥99.8%, Electran for molecular biology
 
undefined
Ethanol absolute ≥99.8%, Electran for molecular biology
 

 

  • Environmentally Preferable:

 

 

  • Assay:
    Min. 99.8 %
  • Appearance:
    Clear colourless liquid
  • DNases (exo- and endonucleases):
    Not detected
  • RNases:
    Not detected
  • Proteases:
    Not detected
  • Acidity or alkalinity:
    Max. 0.0002 meq/g
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Non-volatile residue:
    Max. 0.0010 %
  • Water:
    Max. 0.2 %

 

Frequently Bought Together