Order Entry
United Kingdom
ContactUsLinkComponent
Nitric acid 200 g/l
Nitric acid 200 g/l
Catalog # PRLS87264.290
Supplier:  VWR Collection
CAS Number:  
Nitric acid 200 g/l
Catalog # PRLS87264.290
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
Boiling Pt:∼86 °C (1013 hPa)
EINECS:231-714-2
UN:2031
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Melting Pt:–42 °C
Density:1.2…1.4 g/cm³ (20 °C)
ADR:8 II
Merck Index:13,06610