Methoxyacetyl chloride ≥97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:38870-89-2
:
Formula:C₃H₅ClO₂
MDL Number:MFCD00000728
Boiling Pt:112…113 °C
Density:1.185
:~ 0.3 %
Molecular Weight:108.52 g/mol
Melting Pt:< –40 °C
Flash Pt:28 °C (82 °F)
Storage Temperature:Ambient
EINECS:254-169-2
UN:2920
ADR:8 II
:GHS06
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together