2-Bromo-5-methoxypyridine
Supplier: COMBI-BLOCKS
Certificates
About this item
CAS Number:105170-27-2
:
Formula:C₆H₆BrNO
MDL Number:MFCD07781221
Molecular Weight:188.02 g/mol
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together