Desiccators
Supplier: Brand
Certificates
About this item
Borosilicate glass 3.3 with precisely ground flat flanges.
- Base ring surface is studded to improve stability and help prevent cracks
- Interchangeable lid
Desiccators with sockets are supplied with 24/29 socket and interchangeable stopcock.
Certifications: DIN 12491
Frequently Bought Together