Order Entry
United Kingdom
ContactUsLinkComponent
Calcium chloride hexahydrate 97+%, extra pure
Calcium chloride hexahydrate 97+%, extra pure
Certificates
undefined
Calcium chloride hexahydrate 97+%, extra pure

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless Chunks
  • Titration Complexometric:
    97.0 to 103.0 %
  • Alkalinity:
    ≤0.03 % (as Ca(OH)2)
  • Phosphate (PO4):
    ≤0.003 %
  • Sulfate (SO4):
    ≤0.05 %
  • Iron (Fe):
    ≤0.005 %
  • Acidity (HCl):
    ≤0.025 %
  • Insoluble matter:
    ≤0.025 % (in water and ppt by NH4OH)

Specification Test Results

Related Information

Frequently Bought Together