Sodium dihydrogen phosphate monohydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
CAS Number:10049-21-5
:
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208
Density:2.04 g/cm³ (20 °C)
Molecular Weight:137.99 g/mol
Melting Pt:100
Merck Index:13,08734
Storage Temperature:Ambient
EINECS:231-449-2
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together