Sodium molybdate(VI) dihydrate ≥99.5%, AnalaR NORMAPUR® analytical reagent
: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate
:10102-40-6
:
:Na₂MoO₄·2H₂O
:MFCD00149170
:3.28 g/cm³ (20 °C)
:241.95 g/mol
:687
:13,08718
:Ambient
:231-551-7
- Environmentally Preferable:
- Assay:Min. 99.5 %
- pH (20°C; 5 %):7 - 10
- Cl (Chloride):Max. 50 ppm
- NH4 (Ammonium):Max. 10 ppm
- NO3 (Nitrate):Max. 100 ppm
- PO4 + AsO4 + SiO4 (as PO4):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Fe (Iron):Max. 10 ppm
- Pb (Lead):Max. 10 ppm
Frequently Bought Together