Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
Boiling Pt:>105 °C (1013 hPa)
Melting Pt:323
Density:1.02 g/cm³ (25 °C)
Molecular Weight:40 g/mol
EINECS:215-185-5
ADR:8 III
Merck Index:14,08627
UN:1824
:GHS05
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.2495 - 0.2505 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000)
Packaging Information
spec_VWRC31956.296 spec_VWRC31956.296 spec_VWRC31956.410 spec_VWRC31956.410
Product specification
Frequently Bought Together