Sodium hydroxide 0.111 mol/l (0.111 N) in aqueous solution, AVS TITRINORM volumetric solution, for milk acidity analysis according to Dornic
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
Boiling Pt:1390
Melting Pt:323
Density:1 g/cm³ (25 °C)
Molecular Weight:40 g/mol
Storage Temperature:Ambient
EINECS:215-185-5
ADR:8 III
Merck Index:14,08627
UN:1824
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.1108 - 0.1112 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Frequently Bought Together