Potassium dichromate concentrate for dilution to 1/60 mol in aqueous solution, ConvoL NORMADOSE® volumetric solution
32061.600
:
- Pack type:Plastic ampoule
- Pk:60 mL
- Seq:2385
- Environmentally Preferable:
- Titer (20°C):0.01658 - 0.01675 mol/l
:
:294.18 g/mol
:231-906-6
:3287
:GHS06
:GHS08
:Ambient
:7778-50-9
:K₂Cr₂O₇
:MFCD00011367
:500
:398
:6.1 III
:12,07790
:Dose for preparation of 1 litre of solution.