Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
VWR®, Benzene 200 µg/ml in methanol, organic reference standard
VWR®, Benzene 200 µg/ml in methanol, organic reference standard
123152U
:
VWR Chemicals
:
VWR®, Benzene 200 µg/ml in methanol, organic reference standard
123152U
:
VWR Chemicals
:
Pack type:
Glass ampoule
Pk:
1 mL
CAS No.:
71-43-2
Description:
Benzene 200 µg/ml in methanol
Environmentally Preferable:
Formula:
C₆H₆
Description:
Benzene