Sulphuric acid 10% 10%
Supplier: VWR Collection
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
Boiling Pt:330
Melting Pt:10.38
Density:1.066 g/cm³ (20 °C)
Molecular Weight:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8 II
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together