Sodium hydroxide 30% (w/w) 40% (w/v) in aqueous solution, GPR RECTAPUR® low in nitrogen
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:∼1.33 g/ml (20 °C)
Molecular Weight:40 g/mol
EINECS:215-185-5
ADR:8 II
Merck Index:14,08627
UN:1824
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (W/V):38.6 - 41.7 %
- Appearance:Clear colourless liquid
- Identification (Na):Passes test
- CO3 (as Na2CO3):Max. 0.2 %
- Total N (Nitrogen):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
BSE TSE 191537X-SODIUM HYDROXIDE 40% WV LOW NITROGEN
BSE/TSE Certificates
spec_VWRC191533K spec_VWRC191536N spec_VWRC191537X spec_VWRC19153EB spec_VWRC19153EB
Product specification
More
Frequently Bought Together