Hydrogen peroxide 30% unstabilised, AnalaR NORMAPUR® for trace analysis
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7722-84-1
:
Formula:H₂O₂
MDL Number:MFCD00011333
Boiling Pt:~107 °C (1013 hPa)
Melting Pt:<0 °C
Density:1.11 g/cm³ (20 °C)
Molecular Weight:34.01 g/mol
Storage Temperature:Ambient
EINECS:231-765-0
ADR:5.1 (8) II
Merck Index:14,04798
UN:2014
:GHS07
:GHS05
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:30.0 - 32.0 %
- Acidity:Max. 0.0002 meq/g
- Colouration:Max. 10 APHA
- Evaporation residue:Max. 10 ppm
- Total N (Nitrogen):Max. 3 ppm
- Cl (Chloride):Max. 0.5 ppm
- PO4 (Phosphate):Max. 1 ppm
- SO4 (Sulphate):Max. 1 ppm
- As (Arsenic):Max. 0.01 ppm
- Cd (Cadmium):Max. 0.01 ppm
- Cu (Copper):Max. 0.01 ppm
- Fe (Iron):Max. 0.03 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.02 ppm
- Zn (Zinc):Max. 0.03 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Glass bottle Safebreak for liquids DIN 45 (ZPACK13415250) 500 ml Plastic bottle for liquids DIN 45 (ZPACK13960500)
Packaging Information
23615_HYDROGEN PEROXIDE AB.30 % ANALAR NP T.A._BSE_REV0
BSE/TSE Certificates
spec_VWRC23615.248 spec_VWRC23615.248 spec_VWRC23615.261 spec_VWRC23615.261 spec_VWRC23615.421
Product specification
More
Frequently Bought Together