Hydrochloric acid 35%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.18 g/cm³ (20 °C)
Molecular Weight:36.46 g/mol
Melting Pt:–30 °C
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8 II
REACH:01-2119484862-27
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:35 - 38 %
- Appearance:Clear colourless liquid
- Appearance of solution:Passes test
- Free bromine or chlorine:Passes test
- Evaporation residue:Max. 50 ppm
- Free chlorine:Max. 1 ppm
- Heavy metals (as Pb):Max. 1 ppm
- Ignition residue (SO4):Max. 80 ppm
- Br (Bromide):Max. 5 ppm
- SO3 (Sulphite):Max. 10 ppm
- SO4 (Sulphate):Max. 5 ppm
- As (Arsenic):Max. 1 ppm
- Fe (Iron):Max. 2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 25L Plastic drum for liquids DIN 61 (ZPACK13952503) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC20248.290 spec_VWRC20248.290 spec_VWRC20248.295 spec_VWRC20248.330 spec_VWRC20248.330
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together