Order Entry
United Kingdom
ContactUsLinkComponent
Ethylamine 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Ethylamine 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ethylamine 70% ≥70% in aqueous solution for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Boiling Point:
    39 °C (1013 hPa)
  • Vapor Pressure:
    460 hPa (20 °C)
  • Flash Point:
    -26 °C
  • Explosion Limit:
    3.0 - 12.8 %(V)
  • Density:
    0.81 g/cm3 (20 °C)
  • pH:
    14 (H2O, 20 °C)
  • Ignition Point:
    375 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

Related Information

Frequently Bought Together