Insulin syringes, BD Micro-Fine™
Supplier: EMBECTA
Certificates
About this item
Micro-Fine™ is a single-use syringe for diabetes. It offers a more comfortable and convenient way to inject insulin with three different volumes.
- Made from surgical stainless steel
- Precision and comfort
- Optimum shape and sharpness of the needle tip and bevel
- Electro polished needle shaft
The white and orange protective cap conforms with sterility and a minimal dead space reduces the risk of air bubbles and prevents waste of insulin.
Frequently Bought Together