Folin-Ciocalteu's reagent 1,9 - 2,1 N for analysis of phenols, suitable for determination of total protein by Lowry method, Sigma-Aldrich®
: Folin & Ciocalteu’s phenol reagent
:
:MFCD00132625
:1.22 g/cm³ (20 °C)
:8 III
:M-Clarity™ quality level = MQ200
:3264
:GHS05
:Room Temperature
- Environmentally Preferable:
- Appearance (Turbidity):Clear
- Appearance (Colour):Faint Yellow to Very Dark Yellow and Faint Green-Yellow to Very Dark Green-Yellow
- Appearance (Form):Solution
- Concentration:1.9 - 2.1 N
- Suitability
Suitable for Use in Lowry Protein Determination:Suitable - Recommended Retest Period:4 years
Frequently Bought Together