DePeX mounting medium
Supplier: Serva
Certificates
About this item
Neutral solution of polystyrene and plasticizers in xylene.
- Colorless viscous liquid
- Refractive index: 1,52 to 1,53
- Storage temperature: 15 to 30 °C
Ideal for use in histology.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together