Order Entry
United Kingdom
ContactUsLinkComponent
Anti-evaporation spheres for water baths
Anti-evaporation spheres for water baths
Catalog # 462-8239
Supplier:  GRANT INSTRUMENTS
undefined
Anti-evaporation spheres for water baths
Catalog # 462-8239
Supplier:  GRANT INSTRUMENTS
Specifications
  • Ø:
    20 mm
  • Description:
    Anti-evaporation spheres
  • Material:
    PP
  • Environmentally Preferable:
  • Pk:
    300
Specifications
About this item
Lightweight, economic PP spheres which form a blanket on the liquid surface. Spheres have 20 mm Ø, 275 spheres cover approximately 1000 cm².
  • Reduce heat loss by 77%, and evaporation by 87%
  • Insulate like a solid, and still allow instant access to the liquid in the bath