Order Entry
United Kingdom
ContactUsLinkComponent
Portable turbidity meters, Turbiquant® 1100
Portable turbidity meters, Turbiquant® 1100
Supplier:  Merck
Certificates
undefined
Portable turbidity meters, Turbiquant® 1100
Supplier:  Merck
Specifications
  • Catalog No:
  • 1.18325.0001
  • 1.18324.0001
  • Range:
  • 0,01 - 1100 NTU
  • 0,01 - 1100 NTU
  • Accuracy:
  • ±2,0% or ± last decimal place NTU (0 - 500) ; ±3,0% of measured value (500 - 1100)
  • ±2,0% or ± last decimal place NTU (0 - 500) ; ±3,0% of measured value (500 - 1100)
  • Calibration:
  • Automatic 1 - 3 points
  • Automatic 1 - 3 points
  • Compliance:
  • ISO 7027 or US EPA 180.0
  • ISO 7027 or US EPA 180.0
  • Measurement units:
  • NTU/FNU
  • NTU/FNU
  • Resolution:
  • 0,01; 0,10; 1,00 (depending on measurement range)
  • 0,01; 0,10; 1,00 (depending on measurement range)
  • Sample volume:
  • 15 ml
  • 15 ml
  • Cuvettes Ø×H:
  • 25×45 mm
  • 25×45 mm
Specifications
Related Information
Calibration standards
Product Information
Frequently Bought Together