Rapid heating laboratory chamber furnaces, RWF
The RWF furnaces have free radiating wire-wound elements and highly efficient low thermal mass insulation, which combine to provide an ideal furnace for light to medium duty laboratory applications where rapid thermal response is vital.
Applications in general industry include, ferrous metal heat treatments; annealing, hardening, tempering, stress relieving, and simulation of a larger industrial process, on a laboratory scale. Applications in coatings manufacturing include, testing coatings for resistance to fire, and longevity testing of finished coatings.
: Supplied as standard with a type 301 PID digital temperature controller with single ramp. Alternatively a choice of multi-segment programmers are available. Note furnaces are supplied without plugs and should be hardwired at position of use, 5 litre models require 230 V one phase, 13 litre models require 230 V 1 phase or can alternatively be supplied to 415 V two phase (of three phase) supply at extra charge, 23 litre units require 230 V one phase or 415 V three phase supply, and can be reconfigured on site to one or three phase 'universal' power supply.
: Note: Maximum continuous operating temperature should be 100 °C below the max. temp. of the model shown above.
Frequently Bought Together